Learn More
Medchemexpress LLC Febuxostat-d7 | 1285539-74-3 | MFCD21363616 | 99.1% | 323.42 g/mol | C16H9D7N2O3S | 5 MG
SDP
Catalog No. 5000221366
Encompass_Preferred
Febuxostat-d7 is a deuterium-labeled analog of febuxostat used as a stable-isotope internal standard for analytical and pharmacokinetic studies. It retains the parent scaffold while providing a defined mass shift for accurate quantitation by LC-MS, GC-MS, and NMR. Supplied as a high-purity research-grade standard (CAS 1285539-74-3; formula C16H9D7N2O3S; MW 323.42).
  • High isotopic labeling for reliable mass differentiation.
  • Suitable as an internal standard for LC-MS, GC-MS, and NMR quantitation.
  • High purity minimizes interference in analytical workflows.
  • Stable under recommended storage conditions for reproducible results.
  • Intended for research use only; not for human consumption.
Catalog No. 50-002-21366 Supplier Medchemexpress LLC Supplier No. HY14268S15MG
Febuxostat-d7 is a deuterium-labeled analog of febuxostat used as a stable-isotope internal standard for analytical and pharmacokinetic studies. It retains the parent scaffold while providing a defined mass shift for accurate quantitation by LC-MS, GC-MS, and NMR. Supplied as a high-purity research-grade standard (CAS 1285539-74-3; formula C16H9D7N2O3S; MW 323.42).
  • High isotopic labeling for reliable mass differentiation.
  • Suitable as an internal standard for LC-MS, GC-MS, and NMR quantitation.
  • High purity minimizes interference in analytical workflows.
  • Stable under recommended storage conditions for reproducible results.
  • Intended for research use only; not for human consumption.