Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Febuxostat-d9 | 1246819-50-0 | MFCD21363615 | 98.7% | 325.43 g·mol⁻¹ | C16H7D9N2O3S | 5 MG
SDP

Catalog No. 5000221365
Encompass_Preferred

Febuxostat-d9 is a deuterium-labeled analogue of febuxostat intended for analytical and research use as an internal standard in LC-MS and GC-MS assays. It retains the xanthine oxidase inhibitory profile (reported Ki ≈ 0.6 nM) and is supplied as a purified, isotopically labeled reference material for quantitation, tracer studies, and pharmacokinetic analyses.

  • Deuterium-labeled internal standard for LC/GC-MS quantification.
  • High isotopic enrichment (d9) for clear mass separation.
  • High purity (98.7%) suitable for analytical use.
  • Molecular weight 325.43 g·mol⁻¹ and formula C16H7D9N2O3S.
  • Provided as a 5 mg solid for laboratory assays.
  • CAS 1246819-50-0 for unambiguous identification.

Catalog No. 50-002-21365 Supplier Medchemexpress LLC Supplier No. HY14268S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Febuxostat-d9 is a deuterium-labeled analogue of febuxostat intended for analytical and research use as an internal standard in LC-MS and GC-MS assays. It retains the xanthine oxidase inhibitory profile (reported Ki ≈ 0.6 nM) and is supplied as a purified, isotopically labeled reference material for quantitation, tracer studies, and pharmacokinetic analyses.

  • Deuterium-labeled internal standard for LC/GC-MS quantification.
  • High isotopic enrichment (d9) for clear mass separation.
  • High purity (98.7%) suitable for analytical use.
  • Molecular weight 325.43 g·mol⁻¹ and formula C16H7D9N2O3S.
  • Provided as a 5 mg solid for laboratory assays.
  • CAS 1246819-50-0 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.