Learn More
REVIVAL ANIMAL HEALTH FECAL SOLUTION FECA-MED 1GAL
Catalog No. NC1396268
Encompass
his fecal solution is a standardized solution of sodium nitrate Can be used for all flotation techniques of fecal analysis Ready-to-use solution Pre-mixed for your convenience 1G
Catalog No. NC1396268 Supplier REVIVAL ANIMAL HEALTH Supplier No. 22619801
his fecal solution is a standardized solution of sodium nitrate Can be used for all flotation techniques of fecal analysis Ready-to-use solution Pre-mixed for your convenience 1G