Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copan Diagnostics FecalSwab™ Cary-Blair Collection and Transport System
SDP

Catalog No. 23600972 Shop All Copan Diagnostics Inc. Products
Change view
Click to view available options
Quantity:
50/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-600-972 50/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-600-972 Supplier Copan Diagnostics Supplier No. 4C028S
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Convenient fecal swab and rectal swab collection system converts solid or semi-solid specimens into liquid form for enteric diagnostics. Copan Diagnostics FecalSwab is a convenient system for transporting fecal samples in small instrument-ready tubes saving space and making it easier to transport to the laboratory.

  • FecalSwab is a convenient system for transporting fecal samples in small instrument-ready tubes saving space and making it easier to transport to the laboratory
  • The fecal swab simplifies and standardizes stool sample collection, transport and processing by converting solid or semi-solid specimens into liquid phase, in instrument ready tubes, to facilitate automated fecal sample processing
  • After conducting the primary culture of FecalSwab, the Flocked Swab and cap assembly can be transferred to COPAN Selenite Broth tubes for selective enrichment of Salmonella. The remaining sample can also be used for a molecular enteric diagnostics
  • Each collection kit consists of a package containing a plastic screw-cap tube with conical shaped bottom filled with 2 ml of FecalSwab transport and preservation medium and a specimen collection swab that has a tip flocked with soft nylon fiber

Specifications

Product Type Fecal and Rectal Swab Collection System
Format Liquid
Media Type Cary-Blair
Sterility Sterile
Swab Configuration Single Applicator
Closure Color Orange
Closure Type Screw Cap
Description Regular flocked swab
 with stopper
For Use With (Application) Fecal swab and rectal swab collection
Handle Material Nylon
Includes Screw cap tube with 2mL Cary-Blair medium, One regular flocked swab with stopper
Packaging Sterile pouch
Quantity 50/Pk.
Storage Requirements 5°C to 25°C
Tip Material Nylon Fiber
Tip Type Flocked
Tube Size 12 x 80 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.