Learn More
Med Vet International Fecatector Diagnostic, Fecal Float Device Kit, Laboratory Flotation Device, 50 Kits Per Box
Catalog No. 501180365
Encompass_Preferred
A unique, sanitary leak-proof collection device, to be used by the client for obtaining fecal samples from pets. The same container then doubles as the laboratory flotation device. This eliminates the need for technicians to transfer or contact the stool sample. Comes complete withLaboratory instructions card Easy-to-follow client instruction sheets Compact display box for wall-mounted or counter use.
Catalog No. 50-118-0365 Supplier Med Vet International Supplier No. 6062799
A unique, sanitary leak-proof collection device, to be used by the client for obtaining fecal samples from pets. The same container then doubles as the laboratory flotation device. This eliminates the need for technicians to transfer or contact the stool sample. Comes complete withLaboratory instructions card Easy-to-follow client instruction sheets Compact display box for wall-mounted or counter use.