Learn More
Aviva Systems Biology Corp FECH Antibody - N-terminal region (ARP41865_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41865_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FECH. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human FECH. Alternate names: EPP, FCEApplicationIHC, WB. Species reactivity: Human, Rat
Catalog No. 50-014-58024