Learn More
Sigma Aldrich Fecralloy - Iron-Chromium, Fe72.8Cr22Al5Y0.1Zr0.1
Catalog No. 503091645 Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
50-309-1645 1 Ea.
1 options
Catalog No. 50-309-1645 Supplier Sigma Aldrich Supplier No. GF492010061EA
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Fecralloy - Iron-Chromium, Fe72.8Cr22Al5Y0.1Zr0.1
Specifications
Quantity 1 Ea.