Learn More
Andwin Scientific FectoPRO DNA Transfection Kit
SDP
Catalog No. NC0835231
Encompass_Preferred
The FectoPRO Transfection Kit is specifically designed for enhanced Transient Gene Expression (TGE) in suspension CHO and HEK-293 cells in various serum-free media, using low DNA amount.
Catalog No. NC0835231 Supplier Andwin Scientific Supplier No. 10118842
The FectoPRO Transfection Kit is specifically designed for enhanced Transient Gene Expression (TGE) in suspension CHO and HEK-293 cells in various serum-free media, using low DNA amount.