Learn More
BIOLINK LABORATORIES LLC FECTOPRO TRANSFEC REAGENT 10ML
Catalog No. NC2007828
Encompass
+10mL FectoPRO boosterReagent:FectroPRO®Molecule Delivered:DNAApplications:BioproductionCell Types:Suspension CHO, HEK-293, and derivativesNumber of Transfections:1mL of FectoPRO® is sufficient to transfect 800mL to more than 1L of cell cultureStorage:5°C ± 3°C, stable for 1 to 2 years when stored appropriatelyProvided with:FectoPRO® Booster
Catalog No. NC2007828 Supplier BIOLINK LABORATORIES LLC Supplier No. 101000007
+10mL FectoPRO boosterReagent:FectroPRO®Molecule Delivered:DNAApplications:BioproductionCell Types:Suspension CHO, HEK-293, and derivativesNumber of Transfections:1mL of FectoPRO® is sufficient to transfect 800mL to more than 1L of cell cultureStorage:5°C ± 3°C, stable for 1 to 2 years when stored appropriatelyProvided with:FectoPRO® Booster