Learn More
Fine Science Tools Reusable Feeding Needles, 20 Gauge, 30 mm
SDP
Catalog No. NC9582071
Encompass
Product: Reusable Feeding Needle - Straight/30mm/20G/1.25mm Tip Diameter/ Gauge: 20/ Tip Diameter: 1.25mm/ Shape: Straight/ Tip Shape: Barrel/ Alloy: Stainless Steel/ Length: 30mm/ Sterile: No/ Style: Reusable/ Unit
Catalog No. NC9582071 Supplier Fine Science Tools Supplier No. 1806020
Product: Reusable Feeding Needle - Straight/30mm/20G/1.25mm Tip Diameter/ Gauge: 20/ Tip Diameter: 1.25mm/ Shape: Straight/ Tip Shape: Barrel/ Alloy: Stainless Steel/ Length: 30mm/ Sterile: No/ Style: Reusable/ Unit