Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Federal Signal Portable Loud Speakers

Catalog No. 19811389
Encompass
Encompass_Preferred
Change view
Click to view available options
Detection Range:
1320 ft.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detection Range
19-811-389 1320 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-811-389 Supplier Federal Signal Supplier No. A12SA
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-output megaphones allow you to take control of hectic situations

Lightweight, rugged megaphones with high-impact parts and solid-state design. Special noise-canceling microphone improves voice reproduction even over long distances. Easy-to-hold pistol grip. Available in two models.

Transistorized Megaphone
Is ideal for crowd control.

  • Built-in volume control
  • Trigger switch operation
  • Operates on six standard C batteries
  • 10 watts of power for a range of 800 ft.

Dimensions: 13 3/4L x 7 3/4 in. dia. Weight: 5 lb. White/red.

Electronic Voice Gun
For use in emergency situations and hazardous applications where fire may be a threat.

  • Flame-retardant plastic parts
  • Operates on eight standard AA batteries
  • 16 watts peak power for a range of 1320 ft.

Dimensions: 10L x 7 1/4 in. dia. Weight: 4 lb. Red.

Specifications

Battery Type 8 AA
Color Red
Detection Range 1320 ft.
Diameter (English) Outer 7.25 in.
Material Plastic
Product Type Loudspeaker
Weight (Metric) 1.36 kg
Diameter (Metric) Outer 18.4 cm
For Use With (Application) Ideal for Industrial, Fire, Marine, Athletics, Law Enforcement and Many Other Applications, For Use in Emergency Situations and Hazardous Applications where Fire may be a Threat
Height (English) 10.5 in.
Height (Metric) 26.6 cm
Length (English) 10.13 in.
Length (Metric) 25.7 cm
Weight (English) 3 lb.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.