Learn More
Cayman Chemical FedotozIntartrate 10mg
SDP
Catalog No. 502963973
Encompass_Preferred
A κ-opioid agonist; binds to κ-opioid receptors in mouse brain membranes (Ki = 0.16 nM); reduces gastric extension-induced increases in MAP in a rat model of duodenal acidification-induced gastric hypersensitivity at 10 mg/kg; prevents colonic distension-induced inhibition of gastric motility and emptying in dogs when administered at a dose of 50 UG/kg
Catalog No. 50-296-3973 Supplier Cayman Chemical Supplier No. 2218910 MG
A κ-opioid agonist; binds to κ-opioid receptors in mouse brain membranes (Ki = 0.16 nM); reduces gastric extension-induced increases in MAP in a rat model of duodenal acidification-induced gastric hypersensitivity at 10 mg/kg; prevents colonic distension-induced inhibition of gastric motility and emptying in dogs when administered at a dose of 50 UG/kg