Learn More
Medchemexpress LLC HY-10409 200mg , Fedratinib CAS:936091-26-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10409200MG
Encompass_Preferred
HY-10409 200mg Fedratinib CAS: 936091-26-8 Fedratinib (TG-101348) is a potent, selective, ATP-competitive and orally active JAK2 inhibitor with IC50s of 3 nM for both JAK2 and JAK2V617F kinase. Fedratinib shows 35- and 334-fold selectivity over JAK1 and JAK3, respectively. Fedratinib induces cancer cell apoptosis and has the potential for myeloproliferative disorders research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0462