Learn More
Medchemexpress LLC HY-10409A 100mg , Fedratinib (hydrochloride hydrate) CAS:1374744-69-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10409A100MG
Encompass_Preferred
HY-10409A 100mg Fedratinib (hydrochloride hydrate) CAS: 1374744-69-0 Fedratinib hydrochloride hydrate (TG-101348 hydrochloride hydrate) is a potent, selective, ATP-competitive and orally active JAK2 inhibitor with IC50s of 3 nM for both JAK2 and JAK2V617F kinase. Fedratinib hydrochloride hydrate shows 35- and 334-fold selectivity over JAK1 and JAK3, respectively. Fedratinib hydrochloride hydrate induces cancer cell apoptosis and has the potential for myeloproliferative disorders research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9914