Learn More
Medchemexpress LLC Fedratinib (hydrochloride hydrate) | 1374744-69-0 | 99.9% | 615.62 | 1 G
SDP
Catalog No. 5000365180
Encompass_Preferred
Fedratinib hydrochloride hydrate is a potent, selective, ATP-competitive, and orally active inhibitor of JAK2. It exhibits an IC50 of 3 nM for both JAK2 and JAK2V617F kinase and demonstrates significant selectivity over JAK1 (35-fold) and JAK3 (334-fold). This compound has been shown to induce apoptosis in cancer cells and is considered to have potential for research in myeloproliferative disorders.
  • Potent, selective, ATP-competitive, and orally active JAK2 inhibitor.
  • Targets both JAK2 and JAK2V617F kinase effectively at 3 nM IC50.
  • Exhibits high selectivity against JAK1 (35-fold) and JAK3 (334-fold).
  • Induces apoptosis in cancer cells.
  • Shows promise for research in myeloproliferative disorders.
  • Inhibits proliferation of cell lines carrying the JAK2V617F mutation.
  • Reduces STAT5 phosphorylation.
Catalog No. 50-003-65180 Supplier Medchemexpress LLC Supplier No. 1GHY10409A
Fedratinib hydrochloride hydrate is a potent, selective, ATP-competitive, and orally active inhibitor of JAK2. It exhibits an IC50 of 3 nM for both JAK2 and JAK2V617F kinase and demonstrates significant selectivity over JAK1 (35-fold) and JAK3 (334-fold). This compound has been shown to induce apoptosis in cancer cells and is considered to have potential for research in myeloproliferative disorders.
  • Potent, selective, ATP-competitive, and orally active JAK2 inhibitor.
  • Targets both JAK2 and JAK2V617F kinase effectively at 3 nM IC50.
  • Exhibits high selectivity against JAK1 (35-fold) and JAK3 (334-fold).
  • Induces apoptosis in cancer cells.
  • Shows promise for research in myeloproliferative disorders.
  • Inhibits proliferation of cell lines carrying the JAK2V617F mutation.
  • Reduces STAT5 phosphorylation.