Learn More
Medchemexpress LLC Fedratinib hydrochloride hydrate | 1374744-69-0 | 99.9% | 615.62 | 500 MG
SDP
Catalog No. 5000365814
Encompass_Preferred
Fedratinib hydrochloride hydrate is a potent, selective, ATP-competitive, and orally active JAK2 inhibitor. It has IC50s of 3 nM for both JAK2 and JAK2V617F kinase. This compound exhibits 35-fold selectivity over JAK1 and 334-fold selectivity over JAK3. It induces cancer cell apoptosis and is being researched for its potential in treating myeloproliferative disorders.
  • Potent, selective, ATP-competitive, and orally active JAK2 inhibitor
  • Induces cancer cell apoptosis
  • Potential for myeloproliferative disorders research
  • Shows selectivity over JAK1 and JAK3
  • Available in various quantities for research use
  • Comes with data sheet, COA, SDS, and handling instructions
Catalog No. 50-003-65814 Supplier Medchemexpress LLC Supplier No. 500MGHY10409A
Fedratinib hydrochloride hydrate is a potent, selective, ATP-competitive, and orally active JAK2 inhibitor. It has IC50s of 3 nM for both JAK2 and JAK2V617F kinase. This compound exhibits 35-fold selectivity over JAK1 and 334-fold selectivity over JAK3. It induces cancer cell apoptosis and is being researched for its potential in treating myeloproliferative disorders.
  • Potent, selective, ATP-competitive, and orally active JAK2 inhibitor
  • Induces cancer cell apoptosis
  • Potential for myeloproliferative disorders research
  • Shows selectivity over JAK1 and JAK3
  • Available in various quantities for research use
  • Comes with data sheet, COA, SDS, and handling instructions