Learn More
Med Vet International 10Fr Feeding Tube, Non-Sterile, Each
Catalog No. 502522379
Encompass_Preferred
Red Rubber Latex Feeding Tube. Smooth tip, two oval eyes and funnel end. 16" length and radiopaque.
Catalog No. 50-252-2379 Supplier Med Vet International Supplier No. FT10
Red Rubber Latex Feeding Tube. Smooth tip, two oval eyes and funnel end. 16" length and radiopaque.