Learn More
MilliporeSigma™ Feed Water Conductivity Cell
Catalog No. ZEQ7C0NDC Shop All MilliporeSigma Products
Change view
Click to view available options
Type:
Conductivity Cell
Catalog No. Type
ZEQ7C0NDC Conductivity Cell
1 options
Catalog No. ZEQ7C0NDC Supplier MilliporeSigma™ Supplier No. ZEQ7C0NDC
Add to Cart
Add to Cart
An installation accessory that triggers an alarm if the ionic content of feed water is out of specifications to prevent premature exhaustion of IPAK Meta™ and IPAK Quanta™ polishing cartridges.
  • Use to monitor the ionic quality of feed water for Milli-Q™ EQ 7000 Water Purification Systems
  • An alarm triggers if the ionic content of feed water is out of specifications
  • Prevents premature exhaustion of IPAK Meta™ and IPAK Quanta™ polishing cartridges
TRUSTED_SUSTAINABILITY
Specifications
Type Conductivity Cell
For Use With (Application) Preventing Premature Exhaustion of IPAK Meta™ and IPAK Quanta™ Polishing Cartridges
For Use With (Equipment) Milli-Q™ EQ 7000 Water Purification Systems