Learn More
Kuhner Shaker Inc FEEDBEADS GLUCOSE 12 MM 75
Catalog No. NC1413858
Encompass
FeedBeads, Glucose, 12 mm, 75 mg/48h, Qty 25 - Kuhner Feeding Technology provides an innovative tool for efficient high-throughput screening in fed-batch mode. Our diffusion-driven system is built around a variety of cell nutrients embedded in a porous silica matrix and is delivered sterile and ready-to-use. Contact Kuhner Shaker for additional details and for free samples!
Catalog No. NC1413858 Supplier Kuhner Shaker Inc Supplier No. SMFB08001
Add to Cart
Add to Cart
FeedBeads, Glucose, 12 mm, 75 mg/48h, Qty 25 - Kuhner Feeding Technology provides an innovative tool for efficient high-throughput screening in fed-batch mode. Our diffusion-driven system is built around a variety of cell nutrients embedded in a porous silica matrix and is delivered sterile and ready-to-use. Contact Kuhner Shaker for additional details and for free samples!