Learn More
Kuhner Shaker Inc FEEDBEADS GLUCOSE 24 MM QTY
Catalog No. 502570110
Encompass
FeedBeads, Glucose, 24 mm, Qty 20 - Kuhner Feeding Technology provides an innovative tool for efficient high-throughput screening in fed-batch mode. Our diffusion-driven system is built around a variety of cell nutrients embedded in a porous silica matrix and is delivered sterile and ready-to-use. Contact Kuhner Shaker for additional details and for free samples!
Catalog No. 50-257-0110 Supplier Kuhner Shaker Inc Supplier No. SMFB08002
FeedBeads, Glucose, 24 mm, Qty 20 - Kuhner Feeding Technology provides an innovative tool for efficient high-throughput screening in fed-batch mode. Our diffusion-driven system is built around a variety of cell nutrients embedded in a porous silica matrix and is delivered sterile and ready-to-use. Contact Kuhner Shaker for additional details and for free samples!