Learn More
Fritsch Milling And Sizing Inc Feeding Hopper PVC P-1/2 Cl for Metal-Free Crushing
SDP
Catalog No. 501533889
Encompass
Equipment for iron-free pre-crushing:
For the iron-free pre-crushing of medium hard, brittle samples, we recommend the PVC funnel, as well as fixed and movable crushing jaws made of zirconium oxide and lateral support walls made of zirconium oxide or aluminium.
(Please note: Fixed and movable cruhsing jaws made of zirconium oxide and lateral support walls made of zirconium oxide or aluminium are additionally necessary!)
Catalog No. 50-153-3889 Supplier Fritsch Milling And Sizing Inc Supplier No. 01.7410.00
Equipment for iron-free pre-crushing:
For the iron-free pre-crushing of medium hard, brittle samples, we recommend the PVC funnel, as well as fixed and movable crushing jaws made of zirconium oxide and lateral support walls made of zirconium oxide or aluminium.
(Please note: Fixed and movable cruhsing jaws made of zirconium oxide and lateral support walls made of zirconium oxide or aluminium are additionally necessary!)