Learn More
Dyets Inc FEEDING JAR HOLDER SET
Catalog No. NC9650147
Encompass
Flat stainless steel plate with built in holder for a 4 ounce feeding jar. Comes with food follower and lid. Usually ships same day of order.
Catalog No. NC9650147 Supplier Dyets Inc Supplier No. 910018
Flat stainless steel plate with built in holder for a 4 ounce feeding jar. Comes with food follower and lid. Usually ships same day of order.