Learn More
Fine Science Tools Reusable Feeding Needle, 20 G, 2.25 mm, Curved, Round Tip, 38 mm length
SDP
Catalog No. NC1877906
Encompass
With luer lock hubs. Ideal for transcardiac perfusions with fixative.
Catalog No. NC1877906 Supplier Fine Science Tools Supplier No. 1806520
With luer lock hubs. Ideal for transcardiac perfusions with fixative.