Learn More
Roboz Surgical FEEDING NEEDLES 20G 3' CURVED
SDP
Catalog No. NC9392012
Encompass
Reusable Feeding Needle; Stainless Steel; Curved; 20 Gauge; 3 in. Length; 2.25mm Ball Diameter
Catalog No. NC9392012 Supplier Roboz Surgical Supplier No. FN7911
Reusable Feeding Needle; Stainless Steel; Curved; 20 Gauge; 3 in. Length; 2.25mm Ball Diameter