Learn More
Med Vet International Feeding Needles - Curved 20g x 3", J0194CAG, Each
Catalog No. 5000581946
Encompass_Preferred
Animal Feeding Needle. Luerlock hub with stainless steel tube and nickel-plated silver ball on the end. 20g 3" L
Catalog No. 50-005-81946 Supplier Med Vet International Supplier No. JOR194CAG
Animal Feeding Needle. Luerlock hub with stainless steel tube and nickel-plated silver ball on the end. 20g 3" L