Learn More
SAI Infusion Technologies Rodent Oral Gavage (Feeding) Needle - 17G 85mm with Stainless Steel Bite-Guard
SDP
Numéro de catalogue. NC1509677
Encompass
17G 85mm Flexible and Disposable Feeding Needles with a Stainless Steel Bite-Guard for maximum durability. SAI's smooth needle tip and flexible PTFE tubing prevent damage to the esophagus during dosing, while the unique Bite-Guard prevents any biting damage to the needle. These needles are sterile and packed 5/pk, 20 packs/box. Product is for laboratory animal research only.
Numéro de catalogue. NC1509677 Fournisseur SAI Infusion Technologies Code fournisseur FN1785M
Ajouter au panier
Ajouter au panier
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
17G 85mm Flexible and Disposable Feeding Needles with a Stainless Steel Bite-Guard for maximum durability. SAI's smooth needle tip and flexible PTFE tubing prevent damage to the esophagus during dosing, while the unique Bite-Guard prevents any biting damage to the needle. These needles are sterile and packed 5/pk, 20 packs/box. Product is for laboratory animal research only.
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.