Learn More
SAI Infusion Technologies Rodent Oral Gavage (Feeding) Needle - 20G 38mm
SDP
Catalog No. NC1525820
Encompass
20G 38mm Flexible and Disposable Feeding Needles. SAI's smooth needle tip and flexible PTFE tubing prevent damage to the esophagus during dosing. These needles are sterile and packed 5/pk, 20 packs/box. Product is for laboratory animal research only.
Catalog No. NC1525820 Supplier SAI Infusion Technologies Supplier No. FN2038
 Restricted Item
Add to Cart
Add to Cart
20G 38mm Flexible and Disposable Feeding Needles. SAI's smooth needle tip and flexible PTFE tubing prevent damage to the esophagus during dosing. These needles are sterile and packed 5/pk, 20 packs/box. Product is for laboratory animal research only.