Learn More
Kuhner Shaker Inc FEEDING TECHNOY FOR FED-BATCH
Catalog No. NC2187274
Encompass
FeedBeads, Glycerol, 12 mm, 28 mg/48h, Qty 25 - Kuhner Feeding Technology provides an innovative tool for efficient high-throughput screening in fed-batch mode. Our diffusion-driven system is built around a variety of cell nutrients embedded in a porous silica matrix and is delivered sterile and ready-to-use. Contact Kuhner Shaker for additional details and for free samples!
This item is not returnable. View return policy
Catalog No. NC2187274 Supplier Kuhner Shaker Inc Supplier No. SMFB11001
FeedBeads, Glycerol, 12 mm, 28 mg/48h, Qty 25 - Kuhner Feeding Technology provides an innovative tool for efficient high-throughput screening in fed-batch mode. Our diffusion-driven system is built around a variety of cell nutrients embedded in a porous silica matrix and is delivered sterile and ready-to-use. Contact Kuhner Shaker for additional details and for free samples!