Learn More
Med Vet International 20Fr Feeding Tube, 16", Non-Sterile, Each
Catalog No. 502831708
Encompass_Preferred
Red Rubber Latex Feeding Tube. Smooth tip, two oval eyes and funnel end. 16" length and radiopaque.
Catalog No. 50-283-1708 Supplier Med Vet International Supplier No. FT20
Red Rubber Latex Feeding Tube. Smooth tip, two oval eyes and funnel end. 16" length and radiopaque.