Learn More
Kuhner Shaker Inc FEEDPLATE 48 WELL ROUND
Catalog No. 502570102
Encompass
FeedPlate+, 48 Well Round, Glucose, 36 mg/48h - Kuhner Feeding Technology provides an innovative tool for efficient high-throughput screening in fed-batch mode. Our diffusion-driven system is built around a variety of cell nutrients embedded in a porous silica matrix and is delivered sterile and ready-to-use. Contact Kuhner Shaker for additional details and for free samples!
Catalog No. 50-257-0102 Supplier Kuhner Shaker Inc Supplier No. SMFP02704
FeedPlate+, 48 Well Round, Glucose, 36 mg/48h - Kuhner Feeding Technology provides an innovative tool for efficient high-throughput screening in fed-batch mode. Our diffusion-driven system is built around a variety of cell nutrients embedded in a porous silica matrix and is delivered sterile and ready-to-use. Contact Kuhner Shaker for additional details and for free samples!