Learn More
Kuhner Shaker Inc FEEDPLATE 96 WELL SQUARE
Catalog No. 502570104
Encompass
FeedPlate+, 96 Well Square, Glucose, 18mg/48h - Kuhner Feeding Technology provides an innovative tool for efficient high-throughput screening in fed-batch mode. Our diffusion-driven system is built around a variety of cell nutrients embedded in a porous silica matrix and is delivered sterile and ready-to-use. Contact Kuhner Shaker for additional details and for free samples!
Catalog No. 50-257-0104 Supplier Kuhner Shaker Inc Supplier No. SMFP02702
FeedPlate+, 96 Well Square, Glucose, 18mg/48h - Kuhner Feeding Technology provides an innovative tool for efficient high-throughput screening in fed-batch mode. Our diffusion-driven system is built around a variety of cell nutrients embedded in a porous silica matrix and is delivered sterile and ready-to-use. Contact Kuhner Shaker for additional details and for free samples!