Learn More
Kuhner Shaker Inc FEEDPLATE- GLUCOSE 24 DEEPWEL
Catalog No. 502570108
Encompass
FeedPlate- Glucose, 24 Deepwell plates, square, 18mg/48hr - Kuhner Feeding Technology provides an innovative tool for efficient high-throughput screening in fed-batch mode. Our diffusion-driven system is built around a variety of cell nutrients embedded in a porous silica matrix and is delivered sterile and ready-to-use. Contact Kuhner Shaker for additional details and for free samples!
Catalog No. 50-257-0108 Supplier Kuhner Shaker Inc Supplier No. SMFP00703
FeedPlate- Glucose, 24 Deepwell plates, square, 18mg/48hr - Kuhner Feeding Technology provides an innovative tool for efficient high-throughput screening in fed-batch mode. Our diffusion-driven system is built around a variety of cell nutrients embedded in a porous silica matrix and is delivered sterile and ready-to-use. Contact Kuhner Shaker for additional details and for free samples!