Learn More
Kuhner Shaker Inc FEEDPLATE GLYCEROL 24 WELL
Catalog No. 502570091
Encompass
FeedPlate, Glycerol, 24 Well, Square, High Release - 40 mg/48h - Kuhner Feeding Technology provides an innovative tool for efficient high-throughput screening in fed-batch mode. Our diffusion-driven system is built around a variety of cell nutrients embedded in a porous silica matrix and is delivered sterile and ready-to-use. Contact Kuhner Shaker for additional details and for free samples!
Catalog No. 50-257-0091 Supplier Kuhner Shaker Inc Supplier No. SMFP12003
FeedPlate, Glycerol, 24 Well, Square, High Release - 40 mg/48h - Kuhner Feeding Technology provides an innovative tool for efficient high-throughput screening in fed-batch mode. Our diffusion-driven system is built around a variety of cell nutrients embedded in a porous silica matrix and is delivered sterile and ready-to-use. Contact Kuhner Shaker for additional details and for free samples!