Learn More
Kuhner Shaker Inc FEEDPLATE GLYCEROL 48 WELL R
Catalog No. 502570090
Encompass
FeedPlate, Glycerol, 48 well Round, 19mg/48h - Kuhner Feeding Technology provides an innovative tool for efficient high-throughput screening in fed-batch mode. Our diffusion-driven system is built around a variety of cell nutrients embedded in a porous silica matrix and is delivered sterile and ready-to-use. Contact Kuhner Shaker for additional details and for free samples!
Catalog No. 50-257-0090 Supplier Kuhner Shaker Inc Supplier No. SMFP12004
FeedPlate, Glycerol, 48 well Round, 19mg/48h - Kuhner Feeding Technology provides an innovative tool for efficient high-throughput screening in fed-batch mode. Our diffusion-driven system is built around a variety of cell nutrients embedded in a porous silica matrix and is delivered sterile and ready-to-use. Contact Kuhner Shaker for additional details and for free samples!