Learn More
Grainger Feeler Gauge Set Dimension Type Metric
Catalog No. 502766343
Encompass_Preferred
The Mitutoyo Feeler Gauge Set is a precision measuring tool for checking small gaps and clearances. It contains 16 metric blades ranging from 0.05mm to 1.00mm in 0.05mm increments. The blades are made from hardened stainless steel with ground and lapped measuring faces. They come in a plastic wallet case for protection and easy storage. This high quality feeler gauge set is well-suited for automotive, industrial, and engineering applications that require precise gap and clearance measurements.
Catalog No. 50-276-6343 Supplier Grainger Supplier No. 16X227
The Mitutoyo Feeler Gauge Set is a precision measuring tool for checking small gaps and clearances. It contains 16 metric blades ranging from 0.05mm to 1.00mm in 0.05mm increments. The blades are made from hardened stainless steel with ground and lapped measuring faces. They come in a plastic wallet case for protection and easy storage. This high quality feeler gauge set is well-suited for automotive, industrial, and engineering applications that require precise gap and clearance measurements.