Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fehling's reagent II for sugars, For determination of sugar, Reag. Ph. Eur., solution II: potassium sodium tartrate, alcaline, Solstice

Catalog No. 60046589 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
6 x 1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-046-589 6 x 1 L
60-006-03 500 mL
60-005-99 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-046-589 Supplier Solstice Supplier No. 360196X1L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

for determination of sugar, Reag. Ph. Eur., solution II: potassium sodium tartrate, alcaline

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fehling’s reagent II for sugars
Name Note For determination of sugar, Reag. Ph. Eur., solution II: potassium sodium tartrate, alcaline
CAS 1310-73-2, 304-59-6, 7732-18-5
Boiling Point 100°C
CAS Min % ≥70.0000%, ≥10.0000%
CAS Max % <90.0000%, <30.0000%
MDL Number MFCD00012467
UN Number UN1824
Density 1.23-1.25 g/mL at 20 °C
Synonym Potassium sodium tartrate solution, alkaline, Alkaline potassium sodium tartrate
Grade Reagent Ph. Eur.
Packaging Poly Bottle
Quantity 6 x 1 L
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.