Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fehling’s Solution A Copper Sulfate Solution, Reagents

Catalog No. 01336496
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-496 1 L
01-336-493 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-336-496 Supplier Reagents Holdings Llc Supplier No. C72020001A
Add to Cart
Add to Cart

Fehling’s Solution A Copper Sulfate Solution, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Fehling’s Solution A Copper Sulfate Solution
Color Blue
CAS 7732-18-5, 7758-99-8
Boiling Point 100°C
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.04
Quantity 1 L
Sterility Non-sterile
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.