Learn More
Bioworld Fehling's Solution A Copper Sulfate, 3.46% (w/v), 1 L
Catalog No. 50255514
Encompass_Preferred
Fehling-Soxhlet Solution A (Copper)For detection of reducing sugars. Use with Fehling's solution B.Appearance: LiquidDensity : 1.038-1.060 g/mL at 20 °C
Catalog No. 50-255-514 Supplier Bioworld Supplier No. 402201092
Fehling-Soxhlet Solution A (Copper)For detection of reducing sugars. Use with Fehling's solution B.Appearance: LiquidDensity : 1.038-1.060 g/mL at 20 °C