Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Fehling's Solution B
SCI_ED

Catalog No. S25314 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25314 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25314

missing translation for 'validMainframeMsg'

Test for glucose

This product is an Educational Grade Chemical — no C of A is available.

Specifications

Chemical Name or Material Fehling's Solution, B
CAS 1310-58-3, 6381-59-5, 7732-18-5
Assay Percent Range 25% w/v
Density 1.24g/mL
Packaging Solution
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.