Learn More
Bioworld Fehling's Solution B Alkaline Solution, 500 mL
Catalog No. 50255516
Encompass_Preferred
Fehling-Soxhlet Solution B (Alkaline)For detection of reducing sugars. Use with Fehling's solution A (copper).Appearance: Clear LiquidHazMat Fee Applies
Catalog No. 50-255-516 Supplier Bioworld Supplier No. 402201101
Fehling-Soxhlet Solution B (Alkaline)For detection of reducing sugars. Use with Fehling's solution A (copper).Appearance: Clear LiquidHazMat Fee Applies