Learn More
Bioworld Fehling's Solution B Alkaline Solution, 1 L
Catalog No. 50255517
Encompass_Preferred
Fehling-Soxhlet Solution B (Alkaline)For detection of reducing sugars. Use with Fehling's solution A (copper).Appearance: Clear LiquidHazMat Fee Applies
Catalog No. 50-255-517 Supplier Bioworld Supplier No. 402201102
Fehling-Soxhlet Solution B (Alkaline)For detection of reducing sugars. Use with Fehling's solution A (copper).Appearance: Clear LiquidHazMat Fee Applies