Learn More
Bioworld Fehling's Solution B Alkaline Solution, 4 L
Catalog No. 50255518
Encompass_Preferred
Fehling-Soxhlet Solution B (Alkaline)For detection of reducing sugars. Use with Fehling's solution A (copper).Appearance: Clear LiquidHazMat Fee Applies
Catalog No. 50-255-518 Supplier Bioworld Supplier No. 402201103
Fehling-Soxhlet Solution B (Alkaline)For detection of reducing sugars. Use with Fehling's solution A (copper).Appearance: Clear LiquidHazMat Fee Applies