Learn More
Medchemexpress LLC HY-B0184 10mg Medchemexpress, Felbamate CAS:25451-15-4 Purity:>98%
SDP
Catalog No. 502082401
Encompass_Preferred
Medchemexpress, HY-B0184 10mg Felbamate CAS:25451-15-4 Felbamate (W-554) is a potent nonsedative anticonvulsant whose clinical effect may be related to the inhibition of N-methyl-D-aspartate (NMDA). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2401 Supplier Medchemexpress LLC Supplier No. HYB018410MG
Medchemexpress, HY-B0184 10mg Felbamate CAS:25451-15-4 Felbamate (W-554) is a potent nonsedative anticonvulsant whose clinical effect may be related to the inhibition of N-methyl-D-aspartate (NMDA). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.