Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Felbamate | 25451-15-4 | 99.5% | 5 MG
SDP

Catalog No. 5000405309
Encompass_Preferred

Felbamate is a potent nonsedative anticonvulsant whose clinical effect may be related to the inhibition of N-methyl-D-aspartate (NMDA). It is an anti-epileptic drug used to treat partial seizures in adults and partial and generalized seizures associated with Lennox-Gastaut syndrome in children. It has a proposed dual mechanism of action, acting as a positive modulator of GABAA receptors and a blocker of NMDA receptors.

  • Potent nonsedative anticonvulsant
  • Inhibits N-methyl-D-aspartate (NMDA)
  • Used for treatment of epilepsy
  • Treats partial and generalized seizures
  • Molecular formula: C11H14N2O4
  • Molecular weight: 238.24
  • Appearance: White to off-white solid

Catalog No. 50-004-05309 Supplier Medchemexpress LLC Supplier No. HYB01845MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Felbamate is a potent nonsedative anticonvulsant whose clinical effect may be related to the inhibition of N-methyl-D-aspartate (NMDA). It is an anti-epileptic drug used to treat partial seizures in adults and partial and generalized seizures associated with Lennox-Gastaut syndrome in children. It has a proposed dual mechanism of action, acting as a positive modulator of GABAA receptors and a blocker of NMDA receptors.

  • Potent nonsedative anticonvulsant
  • Inhibits N-methyl-D-aspartate (NMDA)
  • Used for treatment of epilepsy
  • Treats partial and generalized seizures
  • Molecular formula: C11H14N2O4
  • Molecular weight: 238.24
  • Appearance: White to off-white solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.