Learn More
Cayman Chemical Felbamate-d4 500ug
SDP
Catalog No. 502937296
Encompass_Preferred
An internal standard for the quantification of felbamate by GC- or LC-MS
Catalog No. 50-293-7296 Supplier Cayman Chemical Supplier No. 28489500 UG
An internal standard for the quantification of felbamate by GC- or LC-MS