Learn More
Cayman Chemical Felbamate-d4 5mg
SDP
Catalog No. 502937522
Encompass_Preferred
An internal standard for the quantification of felbamate by GC- or LC-MS
Catalog No. 50-293-7522 Supplier Cayman Chemical Supplier No. 284895 MG
Add to Cart
Add to Cart
An internal standard for the quantification of felbamate by GC- or LC-MS