Learn More
TARGETMOL CHEMICALS INC FELCISETRAG 5MG
SDP
Catalog No. 502580476
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Felcisetrag (TD-8954) is a potent and selective orally active 5-HT4 agonist with gastrointestinal prokinetic properties.Felcisetrag shows high affinity for the human recombinant 5-HT4(c) (h5-HT4(c)) receptor with a pKi of 9.4. purity: 100%
Catalog No. 50-258-0476 Supplier TARGETMOL CHEMICALS INC Supplier No. T289345MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Felcisetrag (TD-8954) is a potent and selective orally active 5-HT4 agonist with gastrointestinal prokinetic properties.Felcisetrag shows high affinity for the human recombinant 5-HT4(c) (h5-HT4(c)) receptor with a pKi of 9.4. purity: 100%