Learn More
Kingfisher Biotech Inc Feline BAFF Recombinant Protein (Yeast, Endotoxin - Free) - 5 ug
SDP
Catalog No. 502533667
Encompass
The Feline BAFF yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Feline BAFF applications are for cell culture, ELISA standard, and Western Blot Control. Feline BAFF yeast-derived recombinant protein can be purchased in multiple sizes. Feline BAFF Specifications: (Molecular Weight: 17.1 kDa) (Amino Acid Sequence: AALDPEDTVI QDCLQLIADS DTPTIRKGAY TFVPWLLSFK RGRALEEKEN KILVKETGYF FIYGQVLYTD NTFAMGHLIQ RKKVHVFGDE LSLVNLFRCI QNMPETLPNN SCYSAGIAKL EEGDELQLAI PREDAKISQD GDGTFFGALK LL (152)) (Gene ID: 101088640). For research use only. Made in the USA
Catalog No. 50-253-3667 Supplier Kingfisher Biotech Inc Supplier No. RP1098F005
The Feline BAFF yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Feline BAFF applications are for cell culture, ELISA standard, and Western Blot Control. Feline BAFF yeast-derived recombinant protein can be purchased in multiple sizes. Feline BAFF Specifications: (Molecular Weight: 17.1 kDa) (Amino Acid Sequence: AALDPEDTVI QDCLQLIADS DTPTIRKGAY TFVPWLLSFK RGRALEEKEN KILVKETGYF FIYGQVLYTD NTFAMGHLIQ RKKVHVFGDE LSLVNLFRCI QNMPETLPNN SCYSAGIAKL EEGDELQLAI PREDAKISQD GDGTFFGALK LL (152)) (Gene ID: 101088640). For research use only. Made in the USA