Learn More
ATCC Feline calicivirus; Strain F-9
Catalog No. 502383498
Encompass
Feline calicivirus strain F-9 is propagated in CRFK cells (ATCC CCL-94). This strain was isolated from the upper respiratory tract of a cat.
Catalog No. 50-238-3498 Supplier ATCC Supplier No. VR782
Add to Cart
Add to Cart
Feline calicivirus strain F-9 is propagated in CRFK cells (ATCC CCL-94). This strain was isolated from the upper respiratory tract of a cat.