Learn More
Kingfisher Biotech Inc Feline CCL17 Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1990F025
Encompass
The Feline CCL17 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Feline CCL17 applications are for cell culture, ELISA standard, and Western Blot Control. Feline CCL17 yeast-derived recombinant protein can be purchased in multiple sizes. Feline CCL17 Specifications: (Molecular Weight: 16.9 kDa) (Amino Acid Sequence: VLSGALCFRM KDAELKVLYL HDNQLLAGRL HAGNIIQGEE ISVVPNRSLD AKLSPVILGV QGGSQCLSCG TGQEPTLKLE PVNIMELYLG AKESKSFTFY RRDTGLTSSF ESAAYPGWFL CTVPEADQPL RLTQLSEDAS WDSPITDFYF QQCD (154)) (Gene ID: 101097230). For research use only. Made in the USA
Catalog No. 50-253-2559